Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA4 Monoclonal Antibody (7.2), FITC
GREENER_CHOICE

Catalog No. 501137046
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7046 Supplier Invitrogen™ Supplier No. A15433
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITGA4 Monoclonal Antibody for Flow

The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA4, CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone 7.2
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene ITGA4
Gene Accession No. P13612
Gene Alias 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Mouse
Immunogen Human CD49d.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3676
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.