Learn More
Invitrogen™ ITGA4 Monoclonal Antibody (HP2/1)

Description
Clone HP2/1 binds to both intact and the 80kDa fragment of integrin alpha-4. This antibody can be used in basic studies of VLA-4 mediated adhesion and its interaction with the VCAM-1 structure and has been demonstrated to inhibit cell binding to soluble VCAM-1. This product contains sodium azide; removal by dialysis is recommended prior to use in functional assays. Mouse anti Human CD49d monoclonal antibody, clone HP2/1 recognizes human CD49d also known as integrin alpha-4 or VLA-4 subunit alpha.
Specifications
Specifications
| Antigen | ITGA4 |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | HP2/1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | ITGA4 |
| Gene Accession No. | P13612, Q00651 |
| Gene Alias | 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.