Learn More
Invitrogen™ ITGA5/ITGB1 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA559619

Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term BMS patent anti-Integrin alpha 5 beta 1.
The ITGA5 and ITGB1 genes encode the alpha-5 and beta-1 subunits of integrin receptors, respectively. These integrins are transmembrane receptors that mediate cell adhesion and signaling between the extracellular matrix (ECM) and the cell interior. ITGA5, located on chromosome 12q13, binds with ITGB1, located on chromosome 10p11, to form the alpha 5 beta 1 integrin heterodimer, which specifically interacts with fibronectin, facilitating cell adhesion, migration, proliferation, and survival. Alterations in the expression and functional activity of ITGA5 and ITGB1 have been implicated in various cancers, such as gliomas and non-small cell lung cancer (NSCLC), where they play vital roles in tumor progression, metastasis, and the tumor immune microenvironment. Understanding the roles of these integrins can aid in the development of prognostic biomarkers and therapeutic targets for cancer treatment.
Specifications
ITGA5/ITGB1 Biosimilar | |
Recombinant Monoclonal | |
Unconjugated | |
Human | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
1 mg/mL | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.