Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA5/ITGB1 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA530935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530935 Supplier Invitrogen™ Supplier No. MA530935
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ITGA5 & ITGB1 Heterodimer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA5/ITGB1
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ITGA5
Gene Accession No. P05556, P08648
Gene Alias 4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CD49 antigen-like family member E; Cd49e; ENSMUSG00000051907; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; Fibronectin receptor subunit beta; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; FNRB; glycoprotein IIa; Gm9863; gpIIa; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin beta 1; integrin beta 1 (fibronectin receptor beta); Integrin beta1; integrin beta-1; integrin subunit alpha 5; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); Itga5; Itga5_retired; ITGB1; MDF2; MSK12; very late activation protein 5, alpha subunit; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLA5; VLA-5; VLA5A; VLAB; VLA-BETA
Gene Symbols ITGA5, ITGB1
Host Species Mouse
Immunogen Recombinant Human ITGA5 & ITGB1 Heterodimer protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678, 3688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.