Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735609 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5609 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5609 Supplier Proteintech Group Inc Supplier No. 272241AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrin alpha-5, also known as FNRA or VLA5A, is a protein that in humans is encoded by the ITGA5 gene. The product of this gene belongs to the integrin alpha chain family. Integrins are integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.

Specifications

Antigen ITGA5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ITGA5
Gene Accession No. P08648
Gene Alias CD49e, FNRA, Integrin alpha 5, Integrin alpha F, ITGA5, VLA 5, VLA5A
Gene Symbols ITGA5
Host Species Rabbit
Immunogen ITGA5 Fusion Protein Ag26098
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.