Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA6/ITGB1 Recombinant Rabbit Monoclonal Antibody (312)
GREENER_CHOICE

Catalog No. PIMA530941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530941 Supplier Thermo Scientific Supplier No. MA530941
Add to Cart
Add to Cart

ITGA6/ITGB1 Recombinant Rabbit Monoclonal Antibody (312)

ITGA6/ITGB1 Recombinant Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA6/ITGB1
Applications ELISA
Classification Recombinant Monoclonal
Clone 312
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ITGA6
Gene Accession No. P05556, P23229
Gene Alias Alpha6p; CD29; CD49 antigen-like family member F; CD49f; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; GPIIA; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin beta 1; Integrin beta1; integrin beta-1; integrin subunit alpha 6; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin, alpha 6; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITGA6; ITGA6B; ITGB1; MDF2; MSK12; Processed integrin alpha-6; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLA-6; VLAB; VLA-BETA
Gene Symbols ITGA6, ITGB1
Host Species Rabbit
Immunogen Recombinant Human ITGA6 & ITGB1 Heterodimer protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655, 3688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.