Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA8 Monoclonal Antibody (481709)
GREENER_CHOICE

Catalog No. PIMA524196
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524196 Supplier Invitrogen™ Supplier No. MA524196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA8
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 481709
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ITGA8
Gene Accession No. P53708
Gene Alias AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265
Gene Symbols ITGA8
Host Species Mouse
Immunogen E. coli-derived recombinant human Integrin alpha 8 Glu610-Leu800.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8516
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.