Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGA8 Monoclonal Antibody (481709), PE

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523677
Description
ITGA8 Monoclonal Antibody for Flow
Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
Specifications
ITGA8 | |
Monoclonal | |
PE | |
ITGA8 | |
AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265 | |
Mouse | |
Protein A/G | |
RUO | |
8516 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
481709 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P53708 | |
ITGA8 | |
E. coli-derived recombinant human Integrin alpha 8 Glu610-Leu800. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction