Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547572
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547572 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547572 Supplier Invitrogen™ Supplier No. PA547572
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) Integrin alpha 8 is observed, and less than 1% cross-reactivity with rhIntegrin alpha 5, recombinant mouse (rm) Integrin alpha 5, rhIntegrin alpha V and rmIntegrin alpha V is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ITGA8
Gene Accession No. A2ARA8
Gene Alias AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265
Gene Symbols ITGA8
Host Species Goat
Immunogen CHO-derived recombinant mouse Integrin alpha 8 Phe38-Phe1007.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241226, 364786
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.