Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588950 Supplier Invitrogen™ Supplier No. PA588950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVHVVEFHRQ SPAKILNCTN IECLQISCAV GRLEGGESAV LKVRSRLWAH TFLQRKNDPY ALASLVSFEV KKMPYTDQPA KLPEGSIVIK TSVI; Positive Samples: 293T, LO2, Mouse spleen, Mouse testis, Rat lung; Cellular Location: Cell membrane, Membrane, Single-pass type I membrane protein.

Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.502 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene ITGA8
Gene Accession No. A2ARA8, P53708
Gene Alias AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265
Gene Symbols ITGA8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 907-1000 of human ITGA8 (NP_0036292).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241226, 364786, 8516
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.