Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA9/ITGB1 Monoclonal Antibody (Y9A2)
GREENER_CHOICE

Catalog No. PIMA180920
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180920 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180920 Supplier Invitrogen™ Supplier No. MA180920
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Integrin alpha 9 beta 1 antibody, clone Y9A2 recognizes the human alpha9 integrin, a approximately 140 kDa glycoprotein expressed as a heterodimer in association with the beta1 integrin.

Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA9/ITGB1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Y9A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Itgb1
Gene Accession No. P05556, Q13797, Q9GLP0
Gene Alias ALPHA-RLC; CD29; fibronectin receptor subunit beta; FNRB; glycoprotein IIa; GPIIA; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin beta 1; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin subunit alpha 9; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin, alpha 9; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITGA4L; ITGA9; ITGB1; MDF2; MSK12; RLC; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA
Gene Symbols Itga9, Itgb1
Host Species Mouse
Immunogen alpha9 integrin transfected lambda cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100626707, 100716659, 100719502, 3680, 3688, 397019
Target Species Human, Pig, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.