Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGA9 Polyclonal Antibody, PE

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA546896
Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Integrin alpha 1, alpha 8, alpha 9, alpha L, recombinant mouse Integrin 3, 4, 5, E, and M is observed.
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.
Specifications
ITGA9 | |
Polyclonal | |
Buffered saline solution with BSA and 0.09% sodium azide | |
B8JK39 | |
Itga9 | |
Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979. | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
PE | |
Itga9 | |
(alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin subunit alpha 9; integrin, alpha 9; ITGA4L; Itga9; Itga9l; RLC | |
Goat | |
Antigen affinity chromatography | |
RUO | |
104099 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction