Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAE Mouse anti-Human, Unconjugated, Clone: ITGAE/2474, Abnova™

Catalog No. 89337598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-598 Supplier Abnova Corporation Supplier No. MAB21105
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ITGAE Monoclonal antibody specifically detects ITGAE in Human samples. It is validated for IHC-P.

Specifications

Antigen ITGAE
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ITGAE/2474
Conjugate Unconjugated
Description integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide)
Gene ITGAE
Gene Alias CD103, HUMINAE, MGC141996
Host Species Mouse
Immunogen Recombinant protein corresponding to human ITGAE
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3682
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.