Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAL Mouse anti-Human, FITC, Clone: 4A122, Abnova™

Catalog No. 89934388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-388 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-388 Supplier Abnova Corporation Supplier No. MAB12385
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human ITGAL.

ITGAL encodes the integrin alpha L chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form the integrin lymphocyte function-associated antigen-1 (LFA-1), which is expressed on all leukocytes. LFA-1 plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 (intercellular adhesion molecules 1 through 3), and also functions in lymphocyte costimulatory signaling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen ITGAL
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 4A122
Conjugate FITC
Description Mouse monoclonal antibody raised against human ITGAL.
Dilution Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
Gene Alias CD11A/LFA-1/LFA1A
Gene Symbols ITGAL
Host Species Mouse
Immunogen ITGAL
Purification Method Protein A/G affinity chromatography
Quantity 50 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3683
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.