Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAL Mouse anti-Human, Unconjugated, Clone: DF1524, Abnova™

Catalog No. 89337600 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-600 Supplier Abnova Corporation Supplier No. MAB21107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ITGAL Monoclonal antibody specifically detects ITGAL in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen ITGAL
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone DF1524
Conjugate Unconjugated
Description integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide)
Gene ITGAL
Gene Alias CD11A, LFA-1, LFA1A
Host Species Mouse
Immunogen Human ITGAL
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3683
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.