Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGAV/ITGB6 Monoclonal Antibody (06)
GREENER_CHOICE

Catalog No. PIMA530942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530942 Supplier Invitrogen™ Supplier No. MA530942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ITGAV & ITGB6 Heterodimer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGAV/ITGB6
Applications ELISA
Classification Monoclonal
Clone 06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ITGB6
Gene Accession No. P06756, P18564
Gene Alias 1110004F14Rik; 2210409C20Rik; 2610028E01Rik; 4831415H04Rik; AI1H; alpha v integrin; antigen identified by monoclonal antibody L230; Cd51; D430040G12Rik; Dnmt3l-ps1 pseudogene; integrin alpha V; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin beta 6; integrin beta-6; integrin subunit alpha V; integrin subunit beta 6; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 6; ITGAV; ITGB6; MSK8; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor subunit alpha; VNRA; VTNR
Gene Symbols ITGAV, ITGB6
Host Species Mouse
Immunogen Recombinant Human Human ITGAV & ITGB6 Heterodimer Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685, 3694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.