Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB1, anti-Human, Clone: K20, Abnova™

Catalog No. 89032463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-463 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-463 Supplier Abnova Corporation Supplier No. RAB00120
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen ITGB1
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone K20
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL) Immunohistochemistry (Frozen sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)
Gene Alias CD29/FNRB/GPIIA/MDF2/MSK12/VLA-BETA/VLAB
Gene Symbols ITGB1
Immunogen Original antibody is raised against human T-cell lymphoma cells injected into Biozzi high-responder mice.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3688
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.