Learn More
Invitrogen™ ITGB1 Monoclonal Antibody (4B7R), PE

Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD29 monoclonal antibody, clone 4B7R recognizes the human integrin beta 1 subunit, also known as CD29, a approximately 130 kDa (red) 115 kDa (non-red) single pass type I transmembrane glycoprotein expressed by most leucocytes and mesenchymal stem cells.
Specifications
Specifications
| Antigen | ITGB1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4B7R |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | ITGB1 |
| Gene Accession No. | P05556 |
| Gene Alias | 4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.