Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB1BP2 Monoclonal Antibody (308705)
GREENER_CHOICE

Catalog No. PIMA524025
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524025 Supplier Invitrogen™ Supplier No. MA524025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Integrin beta-1 binding protein 2 (ITGB1BP2), commonly referred to as Melusin, is a gene located on human chromosome 8p21.3. ITGB1BP2 is known for its role in cardiac muscle function, encoding a protein that interacts with integrin β1, which is essential for integrin signaling pathways. This gene is implicated in hypertrophic cardiomyopathy and dilated cardiomyopathy, conditions that affect heart muscle structure and function. ITGB1BP2 gene variations have been observed in families with hypertension or primary hypertrophic cardiomyopathy, although clear genotype/phenotype correlations were not conclusively established. Notably, a specific missense mutation in ITGB1BP2, involving alanine-to-glycine substitution at position 313, has been identified in patients with dilated cardiomyopathy. Melusin, the protein encoded by ITGB1BP2, plays a crucial role in mechanotransduction processes, influencing cellular responses to mechanical stress, which is particularly important in cardiac tissue adaptation and repair mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB1BP2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 308705
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ITGB1BP2
Gene Accession No. Q9UKP3
Gene Alias Chordc3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; Itgb1bp2; Melusin; MGC119214; MSTP015; RGD1565015
Gene Symbols ITGB1BP2
Host Species Mouse
Immunogen E. coli-derived recombinant human Melusin Ser2-Glu347.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.