Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGB1BP2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8013R

Description
ITGB1BP2 may play a role during maturation and/or organization of muscles cells.Specifications
ITGB1BP2 | |
Polyclonal | |
Unconjugated | |
ITGB1BP2 | |
BOS_25863; CHORDC3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; Integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; ITGB1BP2; Melusin; MGC119214; MSTP015; RGD1565015 | |
Rabbit | |
Protein A | |
RUO | |
26548 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UKP3 | |
ITGB1BP2 | |
KLH conjugated synthetic peptide derived from human ITGB1BP2/CHORDC3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction