Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGB1BP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ITGB1BP3 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ITGB1BP3 Polyclonal specifically detects ITGB1BP3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ITGB1BP3 | |
Polyclonal | |
Rabbit | |
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
27231 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VYLDGMKSREELFREVLEDIQNSLLNRSQESAPSPARPARTQGPGRGCGHRTARPAASQQDSM | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EC 2.7.1.22, EC 2.7.1.n4, integrin beta 1 binding protein 3, Integrin beta-1-binding protein 3, MGC126624, MIBPNRK 2, Muscle integrin-binding protein, muscle-specific beta 1 integrin binding protein, nicotinamide riboside kinase 2, Nicotinic acid riboside kinase 2, NmR-K 2, NRK2RNK 2, Ribosylnicotinamide kinase 2, Ribosylnicotinic acid kinase 2 | |
NMRK2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title