Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGB1BP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565897
Description
Immunogen sequence: VYLDGMKSREE LFREVLEDIQ NSLLNRSQES APSPARPART QGPGRGCGHR TARPAASQQD SM Highest antigen sequence identity to the following orthologs - mouse 35%, rat 34%.
ITGB1BP3 catalyzes the phosphorylation of nicotinamide riboside (NR) and nicotinic acid riboside (NaR) to form nicotinamide mononucleotide (NMN) and nicotinic acid mononucleotide (NaMN). The protein reduces laminin matrix deposition and cell adhesion to laminin, but not to fibronectin. It is involved in the regulation of PXN at the protein level and of PXN tyrosine phosphorylation and may play a role in the regulation of terminal myogenesis.
Specifications
ITGB1BP3 | |
Polyclonal | |
Unconjugated | |
NMRK2 | |
2310015C21Rik; integrin beta 1 binding protein 3; integrin beta-1-binding protein 3; ITGB1BP3; MIBP; Muscle integrin-binding protein; muscle-specific beta 1 integrin binding protein; nicotinamide riboside kinase 2; Nicotinic acid riboside kinase 2; nmR-K 2; NMRK2; NRK 2; NRK2; Ribosylnicotinamide kinase 2; ribosylnicotinic acid kinase 2; RNK 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPI5 | |
NMRK2 | |
Recombinant Human ITGB1BP3. Recombinant protein control fragment (Product #RP-106571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction