Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567242 Supplier Invitrogen™ Supplier No. PA567242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VYLDGMKSREE LFREVLEDIQ NSLLNRSQES APSPARPART QGPGRGCGHR TARPAASQQD SM Highest antigen sequence identity to the following orthologs - mouse 35%, rat 34%.

ITGB1BP3 catalyzes the phosphorylation of nicotinamide riboside (NR) and nicotinic acid riboside (NaR) to form nicotinamide mononucleotide (NMN) and nicotinic acid mononucleotide (NaMN). The protein reduces laminin matrix deposition and cell adhesion to laminin, but not to fibronectin. It is involved in the regulation of PXN at the protein level and of PXN tyrosine phosphorylation and may play a role in the regulation of terminal myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB1BP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NMRK2
Gene Accession No. Q9NPI5
Gene Alias 2310015C21Rik; integrin beta 1 binding protein 3; integrin beta-1-binding protein 3; ITGB1BP3; MIBP; Muscle integrin-binding protein; muscle-specific beta 1 integrin binding protein; nicotinamide riboside kinase 2; Nicotinic acid riboside kinase 2; nmR-K 2; NMRK2; NRK 2; NRK2; Ribosylnicotinamide kinase 2; ribosylnicotinic acid kinase 2; RNK 2
Gene Symbols NMRK2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human NMRK2 (Product #RP-106571)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.