Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB2 Mouse anti-Human, Clone: 534CT6.2.1, Abnova™

Catalog No. 89934397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-397 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-397 Supplier Abnova Corporation Supplier No. MAB12320
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ITGB2.

The product of this gene belongs to the integrin beta chain family of proteins. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. This gene encodes the integrin beta chain beta 2. A given chain may combine with multiple partners resulting in different integrins. For example, beta 2 combines with the alpha L chain to form the integrin LFA-1, and combines with the alpha M chain to form the integrin Mac-1. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Defects in this gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]

Specifications

Antigen ITGB2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 534CT6.2.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human ITGB2.
Dilution Immunohistochemistry (1:25) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias CD18/LAD/LCAMB/LFA-1/MAC-1/MF17/MFI7
Gene Symbols ITGB2
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human ITGB2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.