Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB2 Mouse anti-Human, Clone: GRF1 (PE), Abnova™

Catalog No. 89033889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-889 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-889 Supplier Abnova Corporation Supplier No. MAB13824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human ITGB2.

  • High specificity
  • High reactivity

Specifications

Antigen ITGB2
Applications Flow Cytometry
Classification Monoclonal
Clone GRF1
Conjugate PE
Description Mouse monoclonal antibody raised against human ITGB2.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Gene Accession No. P05107
Gene Alias CD18/LAD/LCAMB/LFA-1/MAC-1/MF17/MFI7
Gene Symbols ITGB2
Host Species Mouse
Immunogen Human acute lymphoblastic leukemia (ALL) T cells.
Purification Method Protein A/G purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.