Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB2 Mouse anti-Human, Unconjugated, Clone: 68-5A5, Abnova™

Catalog No. 89337616 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-616 Supplier Abnova Corporation Supplier No. MAB21123
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ITGB2 Monoclonal antibody specifically detects ITGB2 in Human samples. It is validated for IF, Func, Flow Cyt.

Specifications

Antigen ITGB2
Applications Immunofluorescence, Functional Assay, Flow Cytometry
Classification Monoclonal
Clone 68-5A5
Conjugate Unconjugated
Description integrin, beta 2 (complement component 3 receptor 3 and 4 subunit)
Gene ITGB2
Gene Alias CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7
Host Species Mouse
Immunogen Stimulated human leukocytes
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.