Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGB3 Mouse anti-Human, Clone: Y2/51, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14824
Description
Mouse monoclonal antibody raised against native human ITGB3.
- High specificity
- High reactivity
Specifications
ITGB3 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) The optimal working d | |
P05106 | |
ITGB3 | |
Phytohemagglutinin (PHA)-stimulated peripheral blood cells. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
Y2/51 | |
Mouse monoclonal antibody raised against native human ITGB3. | |
In 10 mM PBS. | |
CD61/GP3A/GPIIIa | |
Mouse | |
Protein A/G purification | |
RUO | |
3690 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction