Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB3 Mouse anti-Human, Clone: Y2/51, Abnova™

Catalog No. 89034651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-651 Supplier Abnova Corporation Supplier No. MAB14568
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human ITGB3.

  • High specificity
  • High reactivity

Specifications

Antigen ITGB3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone Y2/51
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human ITGB3.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) The optimal working d
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P05106
Gene Alias CD61/GP3A/GPIIIa
Gene Symbols ITGB3
Host Species Mouse
Immunogen Phytohemagglutinin (PHA)-stimulated peripheral blood cells.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3690
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.