Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB3BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555865 Supplier Invitrogen™ Supplier No. PA555865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVEKLSEEIM EIMQNLSSIQ ALEGSRELEN LIGISCASHF LKREMQKTKE LMTKVNKQKL FEKSTGLPHK ASRHLDSYEF LKAI Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 60%.

ITGB3BP is a transcription coregulator that can have both coactivator and corepressor functions. Isoform 1, but not other isoforms, is involved in the coactivation of nuclear receptors for retinoid X (RXRs) and thyroid hormone (TRs) in a ligand-dependent fashion. ITGB3BP acts as a transcriptional corepressor via its interaction with the NFKB1 NF-kappa-B subunit, possibly by interfering with the transactivation domain of NFKB1. It induces apoptosis in breast cancer cells, but not in other cancer cells, via a caspase-2 mediated pathway that involves mitochondrial membrane permeabilization but does not require other caspases. ITGB3BP may also act as an inhibitor of cyclin A-associated kinase. ITGB3BP may be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB3BP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITGB3BP
Gene Accession No. Q13352
Gene Alias 4930471O16Rik; AU022583; beta 3 endonexin; beta-3-endonexin; beta3-endonexin; Cenpr; CENP-R; Centromere protein R; HSU37139; integrin beta 3 binding protein (beta3-endonexin); integrin beta-3-binding protein; integrin subunit beta 3 binding protein; Itgb3bp; Nrif3; Nuclear receptor-interacting factor 3; TAP20
Gene Symbols ITGB3BP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ITGB3BP. Recombinant protein control fragment (Product #RP-94264).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.