Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGB6 Polyclonal Antibody

ITGB6 Polyclonal Antibody
Supplier: Thermo Scientific PA547309
Description
ITGB6 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
ITGB6 | |
Polyclonal | |
Unconjugated | |
Itgb6 | |
2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
16420, 3694 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P18564, Q9Z0T9 | |
Itgb6 | |
CHO-derived recombinant mouse Integrin beta 6 Gly22-Asn706. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction