Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112585
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112585 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112585 Supplier Invitrogen™ Supplier No. PA5112585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITGB6 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB6
Applications ELISA, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Itgb6
Gene Accession No. P18564
Gene Alias 2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6
Gene Symbols Itgb6
Host Species Rabbit
Immunogen Recombinant Human Integrin beta-6 protein (451-586AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3694
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.