Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549404
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549404 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549404 Supplier Invitrogen™ Supplier No. PA549404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with rabbit based on sequence homology.

ITGB8 (Integrin beta 8) is a member of the integrin beta chain family and is a single-pass type I membrane protein with a VWFA domain and four cysteine-rich repeats. This protein noncovalently binds to an alpha subunit to form a heterodimeric integrin complex. In general, integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ITGB8
Gene Accession No. P26012
Gene Alias 4832412O06Rik; integrin beta 8; Integrin beta8; integrin beta-8; integrin subunit beta 8; integrin, beta 8; ITGB8
Gene Symbols ITGB8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 197-231 amino acids from the Central region of human ITGB8.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.