Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGB8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597884
Description
ITGB8 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ITGB8 (Integrin beta 8) is a member of the integrin beta chain family and is a single-pass type I membrane protein with a VWFA domain and four cysteine-rich repeats. This protein noncovalently binds to an alpha subunit to form a heterodimeric integrin complex. In general, integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| ITGB8 | |
| Polyclonal | |
| Unconjugated | |
| ITGB8 | |
| 4832412O06Rik; integrin beta 8; Integrin beta8; integrin beta-8; integrin subunit beta 8; integrin, beta 8; ITGB8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3696 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.424 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26012 | |
| ITGB8 | |
| Recombinant Human Integrin beta-8 protein (43-320AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction