Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572575
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572575 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572575 Supplier Invitrogen™ Supplier No. PA572575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITGBL1 Polyclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGBL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ITGBL1
Gene Accession No. O95965
Gene Alias B930011D01Rik; integrin beta like 1; integrin beta-like protein 1; integrin subunit beta like 1; integrin, beta-like 1; integrin, beta-like 1 (with EGF-like repeat domains); ITGBL1; OSCP; Osteoblast-specific cysteine-rich protein; ten integrin EGF-like repeat domain-containing protein; TIED; with EGF-like repeat domains
Gene Symbols ITGBL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 334-360 amino acids from the Central region of human ITGBL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9358
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.