Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGBL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109869
Description
Immunogen sequence: CYCGNCYCKA GWHGDKCEFQ CDITPWESKR RCTSPDGKIC SNRGTCVCGE CTCHDVDPTG DWGDIHGDTC ECDERDCRAV YDRYSDDFCS GHGQCNCGRC D.
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
Specifications
| ITGBL1 | |
| Polyclonal | |
| Unconjugated | |
| ITGBL1 | |
| B930011D01Rik; integrin beta like 1; integrin beta-like protein 1; integrin subunit beta like 1; integrin, beta-like 1; integrin, beta-like 1 (with EGF-like repeat domains); ITGBL1; OSCP; Osteoblast-specific cysteine-rich protein; ten integrin EGF-like repeat domain-containing protein; TIED; with EGF-like repeat domains | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 223272, 498564, 9358 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.2 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O95965, Q5PQQ8, Q8VDV0 | |
| ITGBL1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ITGBL1 (NP_004782.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction