Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human/Mouse ITIH2 Synthetic Peptide

Catalog No. pep302
Click to view available options
Quantity:
50 μg

Synthetic Peptide

ITI-HC2 Synthetic Peptide, PEP-302, from Invitrogen™.

The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]). ITI-HC2 may act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes. This peptide corresponds to amino acids E(401) ANNLGLLDPN (411) of human ITI-HC2 and can be used as a blocking peptide with polyclonal antibody PA1-1756. This product cross-reacts with mouse due to sequence homology.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 16425, 3698
Species Human, Mouse
Purification Method purified
Accession Number P19823, Q61703
Common Name Human/Mouse ITIH2
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Lyophilized
Formulation water with no preservative
For Use With (Application) Blocking Assay, Control
Gene Alias ITIH2, HC2, Inter-alpha-trypsin inhibitor heavy chain H2, ITI heavy chain H2, Inter-alpha-inhibitor heavy chain 2, Inter-alpha-trypsin inhibitor complex component II, Serum-derived hyaluronan-associated protein, SHAP
Gene Symbol ITIH2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.