Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITIH1 Polyclonal Antibody

Catalog No. PIPA5121882
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121882 20 μL
PIPA5119211 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121882 Supplier Invitrogen™ Supplier No. PA5121882
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ITIH1 Polyclonal Antibody for Western Blot, IHC (P)

The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ITIH1
Gene Accession No. P19827
Gene Alias CYCSP39; H1P; HCP39; IATIH; IGHEP1; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha trypsin inhibitor, heavy chain 1; inter-alpha-inhibitor heavy chain 1; inter-alpha-trypsin inhibitor complex component III; inter-alpha-trypsin inhibitor heavy chain 1; inter-alpha-trypsin inhibitor heavy chain H1; Intin1; ITI heavy chain H1; ITIH; Itih1; Itih-1; ITI-HC1; MGC126415; Serum-derived hyaluronan-associated protein; SHAP
Gene Symbols ITIH1
Host Species Rabbit
Immunogen Recombinant protein Inter Alpha-Globulin Inhibitor H1. The antigen corresponds to amino acid range 507-819 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.