Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITIH3 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297130H
Description
ITIH3 Polyclonal specifically detects ITIH3 in Human samples. It is validated for ELISA.Specifications
ITIH3 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
3699 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
HRP | |
H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP | |
Produced in rabbits immunized with purified, recombinant ITIH3 (NP_002208.3; Met1-Asp651). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction