Learn More
Invitrogen™ ITIH3 Polyclonal Antibody

Description
ITIH3 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | ITIH3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.79 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ITIH3 |
| Gene Accession No. | Q06033, Q61704, Q63416 |
| Gene Alias | AW108094; H3P; inter-alpha (globulin) inhibitor H3; inter-alpha (globulin) inhibitor, H3 polypeptide; inter-alpha trypsin inhibitor, heavy chain 3; inter-alpha-inhibitor H3 chain; inter-alpha-inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain H3; Intin3; ITI heavy chain H3; ITIH3; Itih-3; ITI-HC3; PAIHC3; pre-alpha (globulin) inhibitor, H3 polypeptide; pre-alpha-inhibitor heavy chain 3; pre-alpha-inhibitor, heavy chain 3; serum-derived hyaluronan-associated protein; SHAP |
| Gene Symbols | ITIH3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.