Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITIH3 Recombinant Rabbit Monoclonal Antibody (049)
GREENER_CHOICE

Catalog No. PIMA530604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530604 Supplier Invitrogen™ Supplier No. MA530604
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ITIH3/ITI-HC3.

This gene encodes the heavy chain subunit of the pre-alpha-trypsin inhibitor complex. This complex may stabilize the extracellular matrix through its ability to bind hyaluronic acid. Polymorphisms of this gene may be associated with increased risk for schizophrenia and major depressive disorder. This gene is present in an inter-alpha-trypsin inhibitor family gene cluster on chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH3
Applications ELISA
Classification Recombinant Monoclonal
Clone 049
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ITIH3
Gene Accession No. Q06033
Gene Alias AW108094; H3P; inter-alpha (globulin) inhibitor H3; inter-alpha (globulin) inhibitor, H3 polypeptide; inter-alpha trypsin inhibitor, heavy chain 3; inter-alpha-inhibitor H3 chain; inter-alpha-inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain H3; Intin3; ITI heavy chain H3; ITIH3; Itih-3; ITI-HC3; PAIHC3; pre-alpha (globulin) inhibitor, H3 polypeptide; pre-alpha-inhibitor heavy chain 3; pre-alpha-inhibitor, heavy chain 3; serum-derived hyaluronan-associated protein; SHAP
Gene Symbols ITIH3
Host Species Rabbit
Immunogen Recombinant Human ITIH3/ITI-HC3 Protein (Met1-Asp651).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.