Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITIH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119213
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119213 200 μL
PIPA5121815 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119213 Supplier Invitrogen™ Supplier No. PA5119213
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody was raised against a recombinant fragment of the protein that is 25 kDa.

This gene encodes a heavy chain component of one of the inter-alpha-trypsin inhibitor (ITI) family members. ITI proteins are involved in extracellular matrix stabilization and in the prevention of tumor metastasis. They are also structurally related plasma serine protease inhibitors and are composed of a light chain and varying numbers of heavy chains. This family member is thought to function as a tumor suppressor in breast and thyroid cancers. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ITIH5
Gene Accession No. Q8BJD1
Gene Alias 4631408O11Rik; 5430408M01Rik; DKFZp686F0145; E130106B02; inter-alpha (globulin) inhibitor H5; inter-alpha trypsin inhibitor heavy chain precursor 5; inter-alpha-inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain family member 5; inter-alpha-trypsin inhibitor heavy chain family, member 5; inter-alpha-trypsin inhibitor heavy chain H5; ITI heavy chain H5; ITIH5; ITI-HC5; KIAA1953; MGC10848; PP14776; UNQ311/PRO354
Gene Symbols ITIH5
Host Species Rabbit
Immunogen Recombinant protein Inter Alpha-Globulin Inhibitor H5. The antigen corresponds to amino acid range 569-737 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209378
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.