Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITIH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551395
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551395 Supplier Invitrogen™ Supplier No. PA551395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLNYLGGLVG ASPWAVFPNY FGGSELVVAG QVQPGKQELG IHLAARGPKD QLLVAHHSEG ATNNSQKAFG CPGEPAPNVA HFIRRLWAYV TIGELLDAHF QARDTTTRHL LAAKVLNLS Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 27%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITIH6
Gene Accession No. Q6UXX5
Gene Alias dJ14O9.1; inter-alpha inhibitor H5-like protein; inter-alpha-trypsin inhibitor heavy chain family member 6; inter-alpha-trypsin inhibitor heavy chain family, member 6; inter-alpha-trypsin inhibitor heavy chain H5-like protein; Inter-alpha-trypsin inhibitor heavy chain H6; ITIH5L; ITIH6; ITI-like protein; UNQ6369; UNQ6369/PRO21074
Gene Symbols ITIH6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ITIH6. Recombinant protein control fragment (Product #RP-91999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347365
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.