Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITK Antibody (SR2174), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32176250UL
Description
ITK Monoclonal antibody specifically detects ITK in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
ITK | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
SR2174 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200 | |
EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
A synthesized peptide derived from human ITK (Uniprot #: Q08881) | |
50 μg | |
Protein Kinase | |
3702 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction