Learn More
Promega ITK Kinase Enzyme System, 1mg
Catalog No. PRV3192 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3192 1 mg
1 options
Catalog No. PRV3192 Supplier Promega Supplier No. V3192
Add to Cart
Add to Cart
Request a Quote
Recombinant human ITK (amino acids 352-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ITK is a member of the TEC family of nonreceptor tyrosine kinases.
  • Easily Screen and Profile ITK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C