Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529668 Supplier Invitrogen™ Supplier No. PA529668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Predicted reactivity: Mouse (95%), Rat (95%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation. The Tec family of non-receptor tyrosine kinases is composed of six proteins designated Tec, Itk (also known as Emt or Tsk), Btk (previously known as Atk, BPK or Emb), Bmx, Txk (also known as Rlk) and Dsrc28C. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain pleckstrin homology (PH) and Tec homology (TH) domains in their amino termini. Itk associates with CD28 and becomes activated subsequent to CD28 ligation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ITK
Gene Accession No. Q08881
Gene Alias EMT; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMT; kinase TLK; LPFS1; LYK; MGC126257; MGC126258; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; Tsk; Tyrosine-protein kinase ITK/TSK; tyrosine-protein kinase LYK
Gene Symbols ITK
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ITK. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3702
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.