Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITK Polyclonal Antibody
ITK Polyclonal Antibody
Supplier: Thermo Scientific PA549363
Description
ITK Polyclonal Antibody for Western Blot

Specifications
ITK | |
Polyclonal | |
Unconjugated | |
ITK | |
EMT; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMT; kinase TLK; LPFS1; LYK; MGC126257; MGC126258; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; Tsk; Tyrosine-protein kinase ITK/TSK; tyrosine-protein kinase LYK | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
16428 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q03526 | |
ITK | |
KLH conjugated synthetic peptide between 156-190 amino acids from the N-terminal region of human Mouse Itk. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction