Learn More
ITL BioMedical SampLok™ Adapter Caps
Catalog No. 21010222 Shop All ITL BioMedical Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
21-010-222 Non-sterile
21-010-223 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-010-222 Supplier ITL BioMedical Supplier No. A100706
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used to collect blood culture samples for bacterial detection testing and laboratory tests
  • Available sterile or non-sterile
  • Allow sampling with variety of blood culture bottles and vacuum test tubes
  • Reduces potential risk of needle stick injury
  • With integrated safety lid
  • Available with or without pre-attached standard slip luer
  • Adapter cap insert available; see cat. no. 21010224
Specifications
For Use With (Application) Allow sampling with variety of blood culture bottles and vacuum test tubes
Sterility Non-sterile
For Use With (Equipment) Adapter Cap with Integrated Safety Lid, No Luer