Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITLN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579543
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SW620 whole cell. IHC: human intestinal cancer tissue. ICC/IF: U20S cell. Flow: U937 cell.
ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.
Specifications
ITLN1 | |
Polyclonal | |
Unconjugated | |
ITLN1 | |
calcium-dependent galactosyl-specific lectin; endothelial lectin HL-1; FLJ20022; Galactofuranose-binding lectin; hIntL; HL1; HL-1; intelectin 1; intelectin 1 (galactofuranose binding); intelectin 1 (galactofuranose binding)-like; intelectin 2; intelectin 5; intelectin a; intelectin-1; Intelectin-1a; intestinal lactoferrin receptor; INTL; Itln; ITLN1; ITLN-1; Itln1a; Itln2; Itln5; Itlna; LFR; omentin; UNQ640/PRO1270 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55600 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q8WWA0 | |
ITLN1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human ITLN1 (19-59aa TDEANTYFKEWTCSSSPSLPRSCKEIKDECPSAFDGLYFLR). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction