Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITM2A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299451PCP
Description
ITM2A Polyclonal antibody specifically detects ITM2A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
ITM2A | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
BRICHOS domain containing 2A, E25ABRICD2A, integral membrane protein 2A, Protein E25 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ITM2A. | |
0.1 mL | |
Stem Cell Markers | |
9452 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction