Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2A Monoclonal Antibody (OTI8C3), OriGene
SDP

Catalog No. 502543915
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3915 Supplier OriGene Supplier No. CF811861
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITM2A Monoclonal antibody specifically detects ITM2A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ITM2A
Applications Western Blot
Classification Monoclonal
Clone OTI8C3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MRC2
Gene Accession No. Q4TU93, Q64449, Q9UBG0
Gene Alias AW108051; BRICD2A; BRICHOS domain containing 2A; E25; E25A; Integral membrane protein 2A; Itm2; ITM2A; Protein E25; UNQ603/PRO1189
Host Species Mouse
Immunogen Full length human recombinant protein of human MRC2 (NP_006030) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17534, 498011, 9902
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.